missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ Protran™ Supported NC Nitrocellulose Membranes: Sheets
Provides exceptional handling characteristics, allowing reprobing
Brand: Cytiva 10600037
This item is not returnable.
View return policy
Description
Individual sheets.
- Membranes consist of nitrocellulose supported by an inert polyester nonwoven material within the membrane
- The support does not affect transfer conditions or results
Specifications
60 cm | |
5/Pk. | |
Nitrocellulose, Inert Polyester |
30 cm | |
Sheet | |
0.2 μm |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- amersham protran supported 0.2µm NC 300mm x 600mm
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva Amersham™ Protran™ Supported NC Nitrocellulose Membranes: Sheets > 0.2um; Sheets; 30 x 60cm; Pk. of 5
Spot an opportunity for improvement?Share a Content Correction