missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Protran™ Supported NC Nitrocellulose Membranes: Sheets

Provides exceptional handling characteristics, allowing reprobing

113.98 € - 482.00 €

Specifications

Format Sheet
Material Nitrocellulose, Inert Polyester
View More Specs

Products 6
Product Code Brand Length (Metric) Width (Metric) Pore Size Quantity Price Quantity & Availability  
Product Code Brand Length (Metric) Width (Metric) Pore Size Quantity Price Quantity & Availability  
15219824
View Documents
Cytiva
10600037
60 cm 30 cm 0.2 μm 5/Pk.
452.00 €
5 sheets
In Stock
to see stock.
Add to basket
 
15239834
View Documents
Cytiva
10600051
20 cm 20 cm 0.45 μm 25/Pk.
482.00 €
25 sheets
In Stock
to see stock.
Add to basket
 
15259834
View Documents
Cytiva
10600053
20 cm 20 cm 0.2 μm 25/Pk.
461.00 €
25 sheets
In Stock
to see stock.
Add to basket
 
15269834
View Documents
Cytiva
10600054
20 cm 20 cm 0.2 μm 10/Pk. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15249834
View Documents
Cytiva
10600052
20 cm 20 cm 0.45 μm 10/Pk. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15209874
View Documents
Cytiva
10600099
9 cm 8 cm 0.2 μm 25/Pk. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Individual sheets.

  • Membranes consist of nitrocellulose supported by an inert polyester nonwoven material within the membrane
  • The support does not affect transfer conditions or results

Specifications

Specifications

Sheet
Nitrocellulose, Inert Polyester
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • amersham protran supported 0.2µm NC 300mm x 600mm

Signal Word
  • Warning

Hazard Category
  • Flammable solid Category 2

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.