missing translation for 'onlineSavingsMsg'
Learn More

Ammonia, ca. 7N solution in methanol, Thermo Scientific Chemicals

Ammonia, H3N, CAS Number-7664-41-7, 67-56-1, am-fol, ammonia, ammonia anhydrous, ammonia, anhydrous, ammonia gas, ammoniak, ammoniakgas, anhydrous ammonia, nitro-sil, spirit of hartshorn, 2.5L, CHEBI:16134, Colorless to Yellow, 6.0 to 7.5N, 0.7700g/mL, 05,15, 14 deg.C, 17.03, GHS Signal Word: Danger

42.61 € - 2324.00 €

Specifications

Chemical Name or Material Ammonia
Color Colorless to Yellow
Name Note ca. 7N Solution in Methanol
CAS 7664-41-7, 67-56-1
Molecular Formula H3N
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10696332
View Documents
Thermo Scientific Acros
133710250
25 mL Glass Bottle
42.61 €
25mL
Please to purchase this item. Need a web account? Register with us today!
10001310
View Documents
Thermo Scientific Acros
133710010
1 L Glass Bottle
198.99 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10761394
View Documents
Thermo Scientific Acros
133710025
2.5 L Glass Bottle
369.84 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10037939
View Documents
Thermo Scientific Acros
133710251
25 L Plastic Can
2324.00 €
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ammonia
ca. 7N Solution in Methanol
H3N
6.0 to 7.5N
05,15
15, 488
QGZKDVFQNNGYKY-UHFFFAOYSA-N
azane
14°C
CHEBI:16134
0.77
Colorless to Yellow
7664-41-7, 67-56-1
NH3
MFCD00011418
0.7700g/mL
ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-fol
N
17.03
222
17.03
Liquid
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonia, ca. 7N solution in methanol, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.