missing translation for 'onlineSavingsMsg'
Learn More
Ammonium carbamate, 98%, Thermo Scientific Chemicals Product Code.: 11343417

Ammonium carbamate, 98%, Thermo Scientific Chemicals

Product Code. 11343417
250 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11343417

Brand: Thermo Scientific Alfa Aesar 018134.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ammoniating agent

Ammonium carbamate is used in the preparation of fertilizers. It acts as an ammoniating agent. It plays an important role in the formation of carbamoyl phosphate, which is essential for urea cycle and pyrimidines production. In addition to this, it is used in the preparation of trimethylsilyl carbamate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1111-78-0
Molecular Formula CH6N2O2
Molecular Weight (g/mol) 78.071
MDL Number MFCD00013010
InChI Key BVCZEBOGSOYJJT-UHFFFAOYSA-N
Synonym ammonium carbamate, ammonium aminoformate, carbamic acid, monoammonium salt, carbamic acid ammonium salt, carbamic acid ammoniate, unii-i2w9615swp, carbamic acid, ammonium salt 1:1, ammoniumcarbamate, anhydride of ammonium carbonate, azanium carbamateShow MoreShow Less
PubChem CID 517232
IUPAC Name azanium;carbamate
SMILES C(=O)(N)[O-].[NH4+]

Specifications

Melting Point 60° (sublimation)
Physical Form Powder
Quantity 250 g
Beilstein 3914091
Solubility Information Soluble in cold water and ammonium hydroxide. Slightly soluble in alcohol. Insoluble in acetone.
Formula Weight 78.07
Density 1.6
Chemical Name or Material Ammonium carbamate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM03L019 Certificate TypeCertificate of Analysis Date21/01/2025 Catalog Number018134.18,018134.30
Lot NumberY21K031 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog Number018134.18,018134.30
Lot NumberU26K009 Certificate TypeCertificate of Analysis Date06/09/2024 Catalog Number018134.18,018134.30
Lot NumberR29K038 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number018134.18,018134.30
Lot NumberR29K033 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number018134.18,018134.30
compliance-icons compliance-icons
Product Identifier
  • Ammonium carbamate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Ammonium carbamate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium carbamate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.