Learn More
Ammonium carbamate, 98%, Thermo Scientific Chemicals
Ammoniating agent
49.86 € - 167.84 €
Chemical Identifiers
CAS | 1111-78-0 |
---|---|
Molecular Formula | CH6N2O2 |
Molecular Weight (g/mol) | 78.071 |
MDL Number | MFCD00013010 |
InChI Key | BVCZEBOGSOYJJT-UHFFFAOYSA-N |
Synonym | ammonium carbamate, ammonium aminoformate, carbamic acid, monoammonium salt, carbamic acid ammonium salt, carbamic acid ammoniate, unii-i2w9615swp, carbamic acid, ammonium salt 1:1, ammoniumcarbamate, anhydride of ammonium carbonate, azanium carbamate |
PubChem CID | 517232 |
IUPAC Name | azanium;carbamate |
SMILES | C(=O)(N)[O-].[NH4+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11333417
|
Thermo Scientific Alfa Aesar
018134.18 |
50 g |
49.86 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11343417
|
Thermo Scientific Alfa Aesar
018134.30 |
250 g |
167.84 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ammonium carbamate is used in the preparation of fertilizers. It acts as an ammoniating agent. It plays an important role in the formation of carbamoyl phosphate, which is essential for urea cycle and pyrimidines production. In addition to this, it is used in the preparation of trimethylsilyl carbamate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1111-78-0 | |
78.071 | |
BVCZEBOGSOYJJT-UHFFFAOYSA-N | |
517232 | |
C(=O)(N)[O-].[NH4+] |
CH6N2O2 | |
MFCD00013010 | |
ammonium carbamate, ammonium aminoformate, carbamic acid, monoammonium salt, carbamic acid ammonium salt, carbamic acid ammoniate, unii-i2w9615swp, carbamic acid, ammonium salt 1:1, ammoniumcarbamate, anhydride of ammonium carbonate, azanium carbamate | |
azanium;carbamate |
Specifications
60° (sublimation) | |
Powder | |
MFCD00013010 | |
ammonium carbamate, ammonium aminoformate, carbamic acid, monoammonium salt, carbamic acid ammonium salt, carbamic acid ammoniate, unii-i2w9615swp, carbamic acid, ammonium salt 1:1, ammoniumcarbamate, anhydride of ammonium carbonate, azanium carbamate | |
BVCZEBOGSOYJJT-UHFFFAOYSA-N | |
azanium;carbamate | |
517232 | |
1.6 |
1111-78-0 | |
CH6N2O2 | |
3914091 | |
Soluble in cold water and ammonium hydroxide. Slightly soluble in alcohol. Insoluble in acetone. | |
C(=O)(N)[O-].[NH4+] | |
78.071 | |
78.07 | |
Ammonium carbamate |
Safety and Handling
P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
H302-H318
missing translation for 'einecsNumber' : 214-185-2
missing translation for 'rtecsNumber' : EY8575000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.