Learn More
STEMCELL Technologies Ammonium Chloride Solution
Reagent for lysis of red blood cells
Brand:  STEMCELL Technologies 07800
Encompass_Preferred
 View more versions of this product
Product Code. 17159311
  • 60.75 € / 100mL
Estimated Shipment: 26-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ammonium Chloride Solution is recommended for the lysis of red blood cells (RBCs) in preparations of human and mouse peripheral blood, spleen, or bone marrow cells
  • It is buffered and optimized for gentle lysis of erythrocytes, with minimal effect on leukocytes
  • It does not contain a fixative agent, therefore leukocytes are viable following RBC lysis
Specifications
Specifications
100 mL
Lysis of Red Blood Cells