Learn More
STEMCELL Technologies Ammonium Chloride Solution
Reagent for lysis of red blood cells
60.75 € - 191.70 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17159311
|
STEMCELL Technologies
07800 |
100 mL |
60.75 €
100mL |
Estimated Shipment: 25-11-2024
Log in to see stock. |
|||||
15570226
|
STEMCELL Technologies
07850 |
500 mL |
191.70 €
500mL |
Estimated Shipment: 25-11-2024
Log in to see stock. |
|||||
Description
- Ammonium Chloride Solution is recommended for the lysis of red blood cells (RBCs) in preparations of human and mouse peripheral blood, spleen, or bone marrow cells
- It is buffered and optimized for gentle lysis of erythrocytes, with minimal effect on leukocytes
- It does not contain a fixative agent, therefore leukocytes are viable following RBC lysis
Specifications
Lysis of Red Blood Cells |