missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Fluoride, ≥98.0% (F), Honeywell™ Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
50 g
Packaging:
Poly bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
CAS | 12125-01-8 |
Color | Colorless |
pH | ∼6 (at 20°C) |
Physical Form | Solid |
Quantity | 50 g |
Molecular Formula | FH4N |
Linear Formula | NH4F |
MDL Number | MFCD00011423 |
UN Number | UN2505 |
InChI Key | LDDQLRUQCUTJBB-UHFFFAOYSA-N |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Safety and Handling


Product Identifier
- Ammonium fluoride
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : ammonium fluoride
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ammonium Fluoride, ≥98.0% (F), Honeywell™ Fluka™ >
Spot an opportunity for improvement?Share a Content Correction