Learn More
Ampicillin Sodium Salt, Ultrapure, Thermo Scientific™
37.47 € - 173.84 €
Chemical Identifiers
CAS | 69-52-3 |
---|---|
Molecular Formula | C16H21N3NaO4S |
Molecular Weight (g/mol) | 374.411 |
MDL Number | MFCD00064313 |
InChI Key | BSFVNXCYXDYHOD-ZQDFAFASSA-N |
Synonym | ampicillin sodium |
PubChem CID | 131673879 |
IUPAC Name | (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium |
SMILES | [HH].CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C.[Na] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15845088
|
Thermo Scientific
J11259.06 |
5 g | Plastic bottle | N/A | |||||
15855088
|
Thermo Scientific
J11259.14 |
25 g | Plastic bottle | N/A | |||||
15865088
|
Thermo Scientific
J11259.22 |
100 g | Plastic bottle | N/A | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
69-52-3 | |
374.411 | |
BSFVNXCYXDYHOD-ZQDFAFASSA-N | |
131673879 | |
[HH].CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C.[Na] |
C16H21N3NaO4S | |
MFCD00064313 | |
ampicillin sodium | |
(2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium |
Specifications
215°C | |
Powder | |
MFCD00064313 | |
ampicillin sodium | |
[HH].CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C.[Na] | |
374.411 | |
371.39 | |
Ampicillin Sodium Salt |
69-52-3 | |
C16H21N3NaO4S | |
4119211 | |
BSFVNXCYXDYHOD-ZQDFAFASSA-N | |
(2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium | |
131673879 | |
Ultrapure |
Safety and Handling
P260-P271-P272-P280g-P285-P302+P352-P304+P340-P314-P333+P313-P342+P311-P363-P501c
H317-H334
missing translation for 'einecsNumber' : 200-708-1
missing translation for 'rtecsNumber' : XH8400000
missing translation for 'tsca' : No
Recommended Storage : Keep cold
Your input is important to us. Please complete this form to provide feedback related to the content on this product.