Learn More
Analtech Glass Thin Layer Chromatography (TLC) Developing Tank
Glass Thin Layer Chromatography (TLC) Developing Tank designed for the development of TLC plates.
168.00 € - 273.00 €
Description
- Tanks are constructed of durable glass and racks are constructed of anodized aluminum.
- Tanks accommodate standard TLC plates.
- The top surface is ground to a precise tolerance for tight sealing to enable tank saturation and optimal plate development.
- Thinline™ tanks accommodate two plates and require less solvent for development.
- Latch-Lid™ units hold lids in place with stainless steel latches.
- Order rack to develop maximum number of plates in a tank.
Specifications
Glass | |
1 Ea. | |
TLC Tank |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Analtech Glass Thin Layer Chromatography (TLC) Developing Tank