Learn More
Laboratoires Anios™ Aniosgel 85 NPC
Laboratoires Anios™ Aniosgel 85 NPC contains no allergenic substances, fragrance, or coloring, and is suitable for hygienic treatment and surgical disinfection of hands
Brand:  Laboratoires Anios™ 1644763
 View more versions of this product
Product Code. 15600115
  • 209.00 € / Pack of 12
Explore more special offers
Description
Description
  • No allergenic substances, fragrance, or coloring
  • Broad antimicrobial spectrum
  • Hygienic treatment in 30 sec.
  • For hygienic treatment and surgical disinfection
  • Active against bacteria, mycobacteria, yeasts, molds, viruses
Specifications
Specifications
Sterilizer/Disinfectant Hand Gel
Ethanol in the presence of thickening, moisturizing and emollient agents, and water.
Pump Bottles
1 L
EN 13727, EN 13727, EN 14348, EN 13624, EN 14476, EN 14476, EN 1500, EN 12791
Hygienic treatment and surgical disinfection of hands
12 Bottles
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Laboratoires Anios™ Aniosgel 85 NPC > 12 x 1L Pump Bottles