missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laboratoires Anios™ Aniosgel 85 NPC
Laboratoires Anios™ Aniosgel 85 NPC contains no allergenic substances, fragrance, or coloring, and is suitable for hygienic treatment and surgical disinfection of hands
154.00 € - 213.00 €
Specifications
Product Type | Sterilizer/Disinfectant Hand Gel |
---|---|
Certifications/Compliance (Europe) | EN 13727, EN 13727, EN 14348, EN 13624, EN 14476, EN 14476, EN 1500, EN 12791 |
Composition | Ethanol in the presence of thickening, moisturizing and emollient agents, and water. |
For Use With (Application) | Hygienic treatment and surgical disinfection of hands |
Quantity | 12 Bottles |
Product Code | Brand | Packaging | Volume (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging | Volume (Metric) | Price | Quantity & Availability | ||||
15680105
|
Laboratoires Anios™
1644748 |
Pump Bottles | 500 mL |
154.00 €
Pack of 12 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15670105
|
Laboratoires Anios™
1644333 |
Airless bottles | 1 L |
213.00 €
Pack of 12 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15600115
|
Laboratoires Anios™
1644763 |
Pump Bottles | 1 L |
209.00 €
Pack of 12 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- No allergenic substances, fragrance, or coloring
- Broad antimicrobial spectrum
- Hygienic treatment in 30 sec.
- For hygienic treatment and surgical disinfection
- Active against bacteria, mycobacteria, yeasts, molds, viruses
Specifications
Sterilizer/Disinfectant Hand Gel | |
Ethanol in the presence of thickening, moisturizing and emollient agents, and water. | |
12 Bottles |
EN 13727, EN 13727, EN 14348, EN 13624, EN 14476, EN 14476, EN 1500, EN 12791 | |
Hygienic treatment and surgical disinfection of hands |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- ANIOSGEL 85 NPC
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P102-Keep out of reach of children.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403-Store in a well-ventilated place.
Supplemental information
- MIXTURE LIST-Contains : ETHYL ALCOHOL, PROPAN-2-OL
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Laboratoires Anios™ Aniosgel 85 NPC