Learn More
Emma Safety Footwear Anne Safety Shoes
Supremely comfortable, sculpted ladies safety shoe. Features SmartTecAir™ technology to remove moisture and perspiration instantly.
Brand:  Emma Safety Footwear MM92251640
 View more versions of this product
Product Code. 15949091
  • 128.00 € / 1 pair
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • S1P-Safety shoe with sole plate for dry work
  • GripForce™ Easy Twist
  • Microfiber construction
  • ESD: Electrostatic Discharge
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver
Specifications
Specifications
EN ISO 20345:2011, EN 12568:2010
Black
Hydro-Tec™ Sanitized Silver
40
Laces
Anne
Microfiber
Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Emma Safety Footwear Anne Safety Shoes > Size: 40