missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Emma Safety Footwear Anne Safety Shoes
Supremely comfortable, sculpted ladies safety shoe. Features SmartTecAir™ technology to remove moisture and perspiration instantly.
103.00 € - 128.00 €
Specifications
Certifications/Compliance | EN ISO 20345:2011, EN 12568:2010 |
---|---|
Closure Type | Laces |
Color | Black |
Design | Anne |
Liner Material | Hydro-Tec™ Sanitized Silver |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
15999081
|
Emma Safety Footwear
MM92251635 |
35 |
128.00 €
1 pair |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||||
15909091
|
Emma Safety Footwear
MM92251636 |
36 | N/A | |||||||
15919091
|
Emma Safety Footwear
MM92251637 |
37 |
128.00 €
1 pair |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||||
15929091
|
Emma Safety Footwear
MM92251638 |
38 |
128.00 €
1 pair |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||||
15939091
|
Emma Safety Footwear
MM92251639 |
39 | N/A | |||||||
15949091
|
Emma Safety Footwear
MM92251640 |
40 |
128.00 €
1 pair |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||||
15959091
|
Emma Safety Footwear
MM92251641 |
41 |
128.00 €
1 pair |
Estimated Shipment: 17-04-2025
Log in to see stock. |
||||||
15969091
|
Emma Safety Footwear
MM92251642 |
42 |
128.00 €
1 pair |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||||
Description
- S1P-Safety shoe with sole plate for dry work
- GripForce™ Easy Twist
- Microfiber construction
- ESD: Electrostatic Discharge
- Chrome-free lining
- Hydro-Tec™ Sanitized Silver
Specifications
EN ISO 20345:2011, EN 12568:2010 | |
Black | |
Hydro-Tec™ Sanitized Silver | |
Steel |
Laces | |
Anne | |
Microfiber |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Emma Safety Footwear Anne Safety Shoes