missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 02-100 Chemical Resistant, 5 Layer Laminated Gloves
Laminated gloves provide extreme resistance against a wide range of chemicals, including biological hazards.
238.00 €
Specifications
Certifications/Compliance (Europe) | CE 0493, EN420:2003 + A1:2009; Category lll |
---|---|
Material | Laminated Film |
Thickness (English) | 2.5 mil |
Thickness (Metric) | 0.06 mm |
EN388 Standard | EN 388:0000 |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
12689781
|
Ansell
2100060 |
6 |
238.00 €
12 pairs |
In Stock
Log in to see stock. ![]() |
||||||
12333609
|
Ansell
2100090 |
9 |
238.00 €
12 pairs |
In Stock
Log in to see stock. ![]() |
||||||
12343609
|
Ansell
2100100 |
10 |
238.00 €
12 pairs |
In Stock
Log in to see stock. ![]() |
||||||
15399622
|
Ansell
2100110 |
11 |
238.00 €
12 pairs |
In Stock
Log in to see stock. |
||||||
Beschreibung
Laminated gloves provide extreme resistance against a wide range of chemicals, including biological hazards.
- Provide flexibility and tactility
- Made from an exceptionally thin chemical-resistant, five-layer laminated film
- Integral, nonwoven liner helps absorb and dissipate perspiration for a cooler, more comfortable fit
- Can be worn as liners under heavier nitrile, neoprene, or PVC gloves
- Hand specific
- Broad spectrum chemical resistance
- Wing-thumb for added wearer comfort
Spezifikation
CE 0493, EN420:2003 + A1:2009; Category lll | |
2.5 mil | |
EN 388:0000 | |
ABCEFGHIMS | |
Smooth | |
AlphaTec™ 02-100 | |
38 cm |
Laminated Film | |
0.06 mm | |
Type A | |
Bacteria, Fungi WITH Virus Protection | |
Unlined | |
15 in. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Ansell AlphaTec™ 02-100 Chemical Resistant, 5 Layer Laminated Gloves