Learn More
Ansell™ Crusader™ Flex Gloves
Non-woven felt insulation and a full-length heat barrier designed for intermittent handling of objects up to 400°F. Ansell™ Crusader™ Flex Gloves can be used in industries such as food service, and glass and metal fabrication.
43.05 €
Products 1
Product Code Brand dimSize Price Quantity & Availability  
Product Code Brand dimSize Price Quantity & Availability  
11733994
View Documents
Ansell™
42-445_9
N/A N/A N/A
Description
Description
  • Front-and-back heat-insulated styles
  • Provides protection from radiant heat and intermittent back-of-hand contact with hot objects
  • Provides excellent grip when handling dry or oily surfaces
  • Gauntlet slip-on cuff
  • Contains no asbestos
  • Length: 10 in. (25.4cm)
Not a replacement for silicate gloves. Designed for intermittent handling of objects up to 400°F (204°C).
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Ansell™ Crusader™ Flex Gloves