missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CD1a; Clone: HI149; BD
Brand: BD Biosciences 563938
379.37 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Host Species: Mouse
Clone: HI149
Isotype: IgG1, κ
Species Reactivity: Hu
Flow cytometry

Specifications
CD1a | |
HI149 | |
Mouse | |
100 Tests | |
Primary |
Monoclonal | |
Brilliant Violet 421 | |
Affinity Purified | |
RUO | |
Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction