missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CD1a; Clone: HI149; BD
282.00 € - 615.00 €
Specifications
Antigen | CD1a |
---|---|
Clone | HI149 |
Classification | Monoclonal |
Conjugate | Brilliant Violet 421 |
Host Species | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15893219
|
BD Biosciences
563939 |
25 Tests |
282.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15883219
|
BD Biosciences
563938 |
100 Tests |
615.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Host Species: Mouse
Clone: HI149
Isotype: IgG1, κ
Species Reactivity: Hu
Flow cytometry
Specifications
CD1a | |
Monoclonal | |
Mouse | |
Human | |
Affinity Purified |
HI149 | |
Brilliant Violet 421 | |
RUO | |
Primary |