Learn More
JAK1 Mouse, Unlabeled, Clone: 73, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610231
216.14 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The JAK family of receptor-associated protein kinases is directly involved in interferon (IFN) response pathways. The JAK family contains at least three members: JAK1, JAK2, and Tyk2. Each protein is approximately 130kDa and contains a C-terminal tyrosine kinase domain, an adjacent kinase or kinase-related domain, and five other domains that are highly conserved among family members. In several human and murine cell lines, JAK1 is rapidly tyrosine phosphorylated in response to IFN-α and IFN-γ. JAK1 is required for the phosphorylation of the transcription factor Stat1 (p91), in response to IFNs-α or IFN-γ. JAK1 is also necessary for the efficient phophorylation of Stat2 (p113) in response to IFN-α and for the phosphorylation of Tyk2 or JAK2 in response to IFNs-α or IFN-γ, respectively.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
JAK1 | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
50 μg | |
Cell Biology | |
Canine, Chicken, Frog, Human, Mouse, Rat | |
IgG2b κ |
Western Blot | |
73 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.