missing translation for 'onlineSavingsMsg'
Learn More

anti-Mouse IgG, peroxidase-linked species-specific whole antibody (from sheep) Secondary Antibody, Cytiva

Highly species-specific anti-Mouse IgG. Cytiva Lifescience™ anti-Mouse IgG, peroxidase-linked species-specific whole antibody (from sheep) Secondary Antibody is designed for applications such as protein blotting, ELISA and immunocytochemistry.

Brand:  Life Science products Cytiva NA931-1ML

497.80 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.



 View more versions of this product

Product Code. 10196124

  • 539.00 € / 1mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Host Species: Sheep
Isotype: IgG
Species Reactivity: Ms

 

TRUSTED_SUSTAINABILITY
Specifications

Specifications

IgG, Whole Molecule
HRP
Affinity Purified
Secondary
Monoclonal
Sheep
1 mL
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : 5-chloro-2-methyl-4-isothiazolin-3-one,2-methyl-2H-isothiazol-3-one
  • UNKNOWN ACUTE-4 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 4% of components with unknown hazards to the aquatic environment.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.