missing translation for 'onlineSavingsMsg'
Learn More

anti-Mouse IgG, peroxidase-linked species-specific whole antibody (from sheep) Secondary Antibody, Cytiva

Highly species-specific anti-Mouse IgG. Cytiva Lifescience™ anti-Mouse IgG, peroxidase-linked species-specific whole antibody (from sheep) Secondary Antibody is designed for applications such as protein blotting, ELISA and immunocytochemistry.

114.96 € - 539.00 €

Specifications

Antigen IgG, Whole Molecule
Classification Monoclonal
Conjugate HRP
Host Species Sheep
Isotype IgG
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10360365
View Documents
Life Science products Cytiva
NA931-100UL
0.1 mL
114.96 €
100µL
Please to purchase this item. Need a web account? Register with us today!
10196124
View Documents
Life Science products Cytiva
NA931-1ML
1 mL
539.00 €
1mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Host Species: Sheep
Isotype: IgG
Species Reactivity: Ms

 

Specifications

Specifications

IgG, Whole Molecule
HRP
IgG
Affinity Purified
Monoclonal
Sheep
Secondary
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains :

SDS
Documents

Documents

Product Certifications