missing translation for 'onlineSavingsMsg'
Learn More

anti-Rabbit IgG, peroxidase-linked species-specific whole antibody (from donkey) Secondary Antibody, Cytiva

A highly species-specific anti-Rabbit IgG. Cytiva Lifescience™ anti-Rabbit IgG, peroxidase-linked species-specific whole antibody (from donkey) Secondary Antibody is used with Amersham™ ECL Western Blotting detection reagents.

Brand:  Life Science products Cytiva NA934-1ML

476.53 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.



 View more versions of this product

Product Code. 10794347

  • 539.00 € / 1mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Host Species: Donkey
  • Isotype: IgG
  • Species Reactivity: Rb
  • Immunogen: Rabbit IgG
  • Applications: Protein Blotting, ELISA, Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IgG
HRP
Affinity Purified
Secondary
Monoclonal
Donkey
1 mL
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : 5-chloro-2-methyl-4-isothiazolin-3-one,2-methyl-2H-isothiazol-3-one
  • UNKNOWN ACUTE-4 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers