missing translation for 'onlineSavingsMsg'
Learn More

anti-Rabbit IgG, peroxidase-linked species-specific whole antibody (from donkey) Secondary Antibody, Cytiva

A highly species-specific anti-Rabbit IgG. Cytiva Lifescience™ anti-Rabbit IgG, peroxidase-linked species-specific whole antibody (from donkey) Secondary Antibody is used with Amersham™ ECL Western Blotting detection reagents.

114.96 € - 539.00 €

Specifications

Antigen IgG
Classification Monoclonal
Conjugate HRP
Host Species Donkey
Isotype IgG
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10379664
View Documents
Life Science products Cytiva
NA934-100UL
0.1 mL
114.96 €
Each
Please to purchase this item. Need a web account? Register with us today!
10794347
View Documents
Life Science products Cytiva
NA934-1ML
1 mL
539.00 €
1mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Host Species: Donkey
  • Isotype: IgG
  • Species Reactivity: Rb
  • Immunogen: Rabbit IgG
  • Applications: Protein Blotting, ELISA, Immunocytochemistry
Specifications

Specifications

IgG
HRP
IgG
Affinity Purified
Monoclonal
Donkey
Secondary
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : 5-chloro2-methyl-4-isothiazolin3-one, 2-methyl-2H-isothiazol3-one

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.