missing translation for 'onlineSavingsMsg'
Learn More
Antimony(III) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11395757

Antimony(III) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11395757
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11395757

Brand: Thermo Scientific Alfa Aesar 043449.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antimony Trichloride is used as a chlorinating agent, as a fireproofing agent in textiles; in bronzing steel and as a mordant in dyeing as well as a caustic in medicine. It is also used as an apoptosis agent that is used to detect cholesterols and carotenoids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-91-9
Molecular Formula Cl3Sb
Molecular Weight (g/mol) 228.11
MDL Number MFCD00011212
InChI Key FAPDDOBMIUGHIN-UHFFFAOYSA-K
Synonym antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimonyShow MoreShow Less
PubChem CID 24814
ChEBI CHEBI:74856
SMILES [Cl-].[Cl-].[Cl-].[Sb+3]

Specifications

Boiling Point 223.5°C
Melting Point 73.4°C
Physical Form Crystals
Quantity 25 g
UN Number UN1733
Merck Index 14,707
Solubility Information It is soluble in alcohol and acetone and acids.
IUPAC Name trichlorostibane
Formula Weight 228.11
Percent Purity 99.999%
Odor Pungent
Packaging Ampule
Sensitivity Moisture sensitive
Density 3.14 g/mL
Assay (metals basis)
Chemical Name or Material Antimony (III) chloride, Ultra dry
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ27K050 Certificate TypeCertificate of Analysis Date31/12/2024 Catalog Number043449.14,043449.22
Lot NumberU06K019 Certificate TypeCertificate of Analysis Date07/10/2024 Catalog Number043449.14,043449.22
Lot NumberP22K012 Certificate TypeCertificate of Analysis Date08/04/2024 Catalog Number043449.14,043449.22
Lot NumberS25G062 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number043449.14,043449.22
Lot NumberJ22L12 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number043449.14,043449.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Antimony(III) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.