missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

94.97 € - 1218.06 €

Chemical Identifiers

CAS 7790-44-5
Molecular Formula I3Sb
Molecular Weight (g/mol) 502.47
MDL Number MFCD00016322
InChI Key KWQLUUQBTAXYCB-UHFFFAOYSA-K
Synonym antimony iii iodide, stibine, triiodo, antimony triiodide, antimon iii iodid, antimony iodide sbi3, antimony iodide, antimony iii iodide, ultra dry, antimony iii triiodide, hsdb 437, triiodostibine #
PubChem CID 24630
IUPAC Name triiodostibane
SMILES [Sb+3].[I-].[I-].[I-]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11330038
SDS View Documents
Thermo Scientific Alfa Aesar
042831.04
2 g
94.97 €
2g
Please to purchase this item. Need a web account? Register with us today!
11340038
SDS View Documents
Thermo Scientific Alfa Aesar
042831.09
10 g
335.67 €
10g
Please to purchase this item. Need a web account? Register with us today!
11320038
SDS View Documents
Thermo Scientific Alfa Aesar
042831.18
50 g
1218.06 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Antimony(III) iodide is used as a dopant in the preparation of thermoelectric materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
Specifications

Chemical Identifiers

7790-44-5
502.47
KWQLUUQBTAXYCB-UHFFFAOYSA-K
24630
[Sb+3].[I-].[I-].[I-]
I3Sb
MFCD00016322
antimony iii iodide, stibine, triiodo, antimony triiodide, antimon iii iodid, antimony iodide sbi3, antimony iodide, antimony iii iodide, ultra dry, antimony iii triiodide, hsdb 437, triiodostibine #
triiodostibane

Specifications

420°C
7790-44-5
I3Sb
UN3260
antimony iii iodide, stibine, triiodo, antimony triiodide, antimon iii iodid, antimony iodide sbi3, antimony iodide, antimony iii iodide, ultra dry, antimony iii triiodide, hsdb 437, triiodostibine #
KWQLUUQBTAXYCB-UHFFFAOYSA-K
triiodostibane
24630
99.999%
Ampule
(metals basis)
168°C
Powder
MFCD00016322
14,710
Soluble in benzene,alcohol,acetone,CS2,HCl,KI,SnCl4,C2H7N. Insoluble in CHCl3,and CCl4.Soluble in benzene,alcohol,acetone,carbondisulfide,hydrochloric acid,potassium iodide,tinterachloride,dimethylamine. Slightly soluble in ethanol and acetone. Insoluble in chloroform and carbonterachloride.
[Sb+3].[I-].[I-].[I-]
502.47
502.46
Acrid
Moisture sensitive
Antimony (III) iodide, Ultra dry
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302+H332

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III

missing translation for 'einecsNumber' : 232-205-8

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Ampouled under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.