Learn More
Antimony(V) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals
82.61 € - 506.52 €
Chemical Identifiers
CAS | 7647-18-9 |
---|---|
Molecular Formula | Cl5Sb |
Molecular Weight (g/mol) | 299.02 |
MDL Number | MFCD00011213 |
InChI Key | VMPVEPPRYRXYNP-UHFFFAOYSA-I |
Synonym | antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid german |
PubChem CID | 24294 |
IUPAC Name | antimony pentachloride |
SMILES | Cl[Sb](Cl)(Cl)(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11393837
|
Thermo Scientific Alfa Aesar
035677.03 |
1 g |
82.61 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11383837
|
Thermo Scientific Alfa Aesar
035677.09 |
10 g |
506.52 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as a polymerization catalyst, a chlorinating agent, an intermediate in chemical synthesis, also a strong oxidizing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7647-18-9 | |
299.02 | |
VMPVEPPRYRXYNP-UHFFFAOYSA-I | |
24294 | |
Cl[Sb](Cl)(Cl)(Cl)Cl |
Cl5Sb | |
MFCD00011213 | |
antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid german | |
antimony pentachloride |
Specifications
79°C (22mmHg) | |
7647-18-9 | |
Cl5Sb | |
UN1730 | |
antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid german | |
VMPVEPPRYRXYNP-UHFFFAOYSA-I | |
antimony pentachloride | |
24294 | |
99.999% | |
1.601 | |
2.336 g/mL | |
Antimony(V) chloride, Ultra dry |
2.8°C | |
Liquid | |
MFCD00011213 | |
14,695 | |
Soluble in water (react),chloroform,hydrochloric acid,and carbon tetrachloride. | |
Cl[Sb](Cl)(Cl)(Cl)Cl | |
299.02 | |
299.02 | |
Pungent | |
Moisture sensitive | |
(metals basis) |
Safety and Handling
GHS H Statement
H314
Causes severe skin burns and eye damage.
P260-P264b-P270-P271-P280-P301+P312-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P363-P501c
H302-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ANTIMONY PENTACHLORIDE, LIQUID
missing translation for 'einecsNumber' : 231-601-8
missing translation for 'rtecsNumber' : CC5075000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only