missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP1G2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17907-1-AP-150UL
This item is not returnable.
View return policy
Specifications
AP1G2 | |
Polyclonal | |
Unconjugated | |
AP1G2 | |
AP1G2, G2AD, Gamma2 adaptin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100362547, 11766, 8906 | |
-20°C | |
Liquid |
Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O75843, O88512 | |
AP1G2 | |
AP1G2 Fusion Protein Ag12300 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |