missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP1G2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | AP1G2 |
---|---|
Concentration | 0.24 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16837914
|
Proteintech
17907-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16827914
|
Proteintech
17907-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
AP1G2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
O75843, O88512 | |
100362547, 11766, 8906 | |
AP1G2 Fusion Protein Ag12300 | |
Primary | |
-20°C | |
AP1G2 |
0.24 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
AP1G2, G2AD, Gamma2 adaptin | |
AP1G2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |