Learn More
Product Code | Brand | Quantity | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Product Type | Price | Quantity & Availability | |||||
12971141
|
Fisherbrand™
800017 |
100 g | M Grease |
257.00 €
100g |
Estimated Shipment: 01-04-2025
Log in to see stock. ![]() |
|||||
12961141
|
Fisherbrand™
800016 |
50 g | L Grease |
485.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12991141
|
Fisherbrand™
800019 |
50 g | AP101 Grease |
248.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Apiezon grease
- For general use in Fisherbrand rotary vane pumps
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Apiezon Grease