missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Apiezon Grease

Lubricate pumps with this grease. Fisherbrand™ Apiezon Grease is designed for use with Fisherbrand™ rotary vane pumps.

Brand:  Fisherbrand™ 800017

 View more versions of this product

Product Code. 12971141

  • 257.00 € / 100g
Estimated Shipment: 07-04-2025
to see stock.
Edge
Explore more special offers
Add to basket

You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy

Description

Description

  • Apiezon grease
  • For general use in Fisherbrand rotary vane pumps
TRUSTED_SUSTAINABILITY
Specifications

Specifications

M Grease
20°C 1.7x10-9 torr
100 g
10°C to 30°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Apiezon Grease > Quantity: 100g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.