missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ APO BrdU (TUNEL) Kit

APO BrdU (TUNEL) Kit

Brand:  Molecular Probes™ KHO1001

Product Code. 10690914

  • 1120.00 € / 60 tests

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The ApoTarget™ APO-BRDU Kit is a two-color staining method for labeling DNA breaks and total cellular DNA to detect apoptotic cells by flow cytometry.

View a selection guide for all apoptosis assays for flow cytometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

60 tests
Apo-BRDU Tunel Kit
488
60
ApoTarget™
Flow Cytometry
FITC (Fluorescein), PI (Propidium Iodide)
Tube
Contains pre-coated plate, standard, detector antibody, diluents, wash buffer, chromogen, stop solution and complete protocol
FITC: 490/525
PI: 535/617
Green, Red
Fluorescence
Flow Cytometer
Classic Dyes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.