missing translation for 'onlineSavingsMsg'
Learn More

SYBR™ Power SYBR™ Green PCR Master Mix

Offers superior sensitivity and reproducibility, detecting as few as two copies of a target gene over a broad range of template concentrations.

Brand:  SYBR™ 4368702

1328.25 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10209284

  • 2415.00 € / 1000 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: SYBR™ Green 1 Dye, AmpliTaq Gold DNA Polymerase LD, dNTPs with dUTP/dTTP blend, Passive Reference 1, and optimized buffer components

Requires : Template and primers
This item is not returnable. View return policy

Description

Description

Get superior nucleic acid quantitation with Applied Biosystems™ Power SYBR™ Green Master Mix.

  • Everything needed for SYBR™ Green dye-based PCR amplification and detection in a convenient single-tube format
  • Power SYBR™ Green PCR Master Mix contains all of the components (excluding the template and primers) for superior SYBR™ Green reagent-based real-time PCR
  • The optimized formulation contains highly purified AmpliTaq Gold™ DNA Polymerase, LD to offer increased sensitivity
  • Contains a blend of dTTP/dUTP that is compatible with AmpErase™ UNG to minimize carryover contamination
  • Includes a proprietary version of ROX™ dye, an internal passive reference, to normalize non-PCR–related fluorescence fluctuations
  • This minimizes well-to-well variability that can result from a variety of causes, such as pipetting error and sample evaporation
  • Power SYBR™ Green reagent-based PCR chemistry easily replaces the existing SYBR™ Green PCR Master Mix in Applied Biosystems™ protocols using the same setup and thermal cycling conditions
  • Store at -20°C

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

The 2X mix contains SYBR™ Green 1 Dye, AmpliTaq Gold™ DNA Polymerase LD, dNTPs with dUTP/dTTP blend, Passive Reference 1, and optimized buffer components. Contains 1 × 50 mL bottle, sufficient for 1000 reactions of 50 μL each.

Store at -20°C.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

Bottle
qPCR
Standard
SYBR Green
SYBR™
LD (Low DNA)
1000 Reactions of 50 μL Each
2X
DNA (Genomic), cDNA
SYBR
High
AmpliTaq Gold DNA Polymerase, AmpliTaq Gold LD
7300 System, 7500 System, 7700 System, 7900HT Fast System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, ViiA™ 7 System
ROX (Pre-mixed)
Real Time PCR SYBR Master Mix
5 x 5 mL
Gene Expression
1000 Reactions
5 x 5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.