missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Platinum™ Multiplex PCR Master Mix

The Platinum Multiplex PCR Master Mix is designed specifically for endpoint multiplex PCR.

201.00 € - 5250.00 €

Specifications

GC-Rich PCR Performance Low
Polymerase Platinum Taq DNA Polymerase
Reaction Speed Standard
Multiplex Capability Up to 20-plex
Shipping Condition Dry Ice
View More Specs

Includes: Platinum Multiplex PCR Master Mix, 2X; GC Enhancer

Products 3
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
12373583
View Documents
Applied Biosystems™
4464268
50 Reactions
201.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
12383583
View Documents
Applied Biosystems™
4464269
250 Reactions
744.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
12027809
View Documents
Applied Biosystems™
4464270
1000 Reactions
5250.00 €
2000 reactions
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Platinum Multiplex PCR Master Mix is a high-specificity, high-throughput solution designed specifically for endpoint multiplex PCR and supports easy multiplexing with minimal optimization.

The performance of the Platinum Multiplex PCR Master Mix over a wide range of amplicon sizes permits the amplification of templates from 50 bp to 2.5 kb, greatly enhancing workflow flexibility. Coupled with its 20-plex capability and absence of primer dimers, it not only provides a high-throughput solution but also boasts high specificity through fewer non-specific primer binding events, resulting in less reaction and primer waste.

Features

  • Amplifies up to 20 amplicons in a single reaction
  • Amplifies products from 50 bp to 2.5 kb
  • Performs multiplex PCR reactions with minimal optimization

Order Info

Shipping Condition: Dry Ice

Specifications

Specifications

Low
Standard
Dry Ice
2X
Built-In Hot Start
2.5 kb or less
Platinum Taq DNA Polymerase
Up to 20-plex
Hot-start PCR, Multiplex PCR
1X
SuperMix or Master Mix
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.