missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Random Hexamers (50 μM)

Random Hexamers are short oligodeoxyribonucleotides of random sequence [d(N)6] that anneal to random complementary sites on a target DNA or RNA to serve as primers for DNA synthesis by a DNA polymerase or reverse transcriptase.

Brand:  Applied Biosystems™ N8080127

Product Code. 10609275

  • 88.50 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Contains Random Hexamers (100 μL) at a concentration of 50 μM. Sufficient for 100 reverse transcriptions of 20 μL each.
This item is not returnable. View return policy

Description

Description

Random Hexamers are short oligodeoxyribonucleotides of random sequence [d(N)6] that anneal to random complementary sites on a target DNA or RNA to serve as primers for DNA synthesis by a DNA polymerase or reverse transcriptase.

For this product five nmoles of primer are supplied at a concentration of 50 μM.

Applications
Most commonly used for complementary DNA (cDNA) synthesis priming, these random hexamers may be used for a variety of applications including:
• Two-step RT-qPCR
• cDNA Synthesis
• RT-PCR

Order Info

Shipping Conditions: Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Primer
Dry Ice
Random Hexamer
DNA Synthesis
• Random Hexamers (100 μL at 50 μM), sufficient for 100 reverse transcriptions of 20 μL each

Store at –15 to –25°C.
50 μM
5 nmol, 100 μL
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3016247 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberN8080127
Lot Number2435466 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberN8080127
Lot Number2500641 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberN8080127
Lot Number2524284 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberN8080127
Lot Number2524302 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberN8080127
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Applied Biosystems™ Random Hexamers (50 μM) > 1 kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.