missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYBR™ Fast SYBR™ Green Master Mix
Designed for speed, SYBR™ Green Master Mix provides a fast, reliable, and cost-effective solution for RT-PCR application without compromising sensitivity, specificity, dynamic range, or PCR efficiency.
Brand: SYBR™ 4385610
Includes: SYBR Green I Dye, AmpliTaq Gold Fast DNA Polymerase LD, dNTPs with dUTP/dTTP blend, Passive Reference I and optimized buffer components
Requires : Template and primers
This item is not returnable. View return policy
Description
- A fast, reliable, and cost-effective solution for real-time PCR applications without compromising sensitivity, specificity, dynamic range, or PCR efficiency
- Real-time PCR results in as fast as 35 minutes
- Capable of detecting very low copies of target
- Minimizes primer-dimer and non-specific amplification
- Consistent amplification across a wide dynamic range
- Contains all of the components, excluding the template and primers, in a convenient 2X master mix
- Store at -5°C to -30°C
Includes the following components in an optimized buffer:
- AmpliTaq™ Fast DNA Polymerase, UP, a highly purified DNA polymerase designed to allow instant hot start
- Minimizes non-specific product formation and enables reactions to be set up at room temperature
- SYBR™ Green I dye to enable detection of double-stranded DNA
- Deoxynucleotides (dNTPs) to help maintain optimal PCR results
- Uracil-DNA Glycosylase (UDG) designed to reduce carryover contamination
- Passive internal reference based on proprietary ROX™ dye to enable increased precision
Order Info
Shipping Condition: Dry Ice
Specifications
1 x 1 mL vial. The 2X mix contains SYBR™ Green 1 Dye, AmpliTaq Gold™ Fast DNA Polymerase LD, dNTPs with dUTP⁄dTTP blend, Passive Reference 1 and optimized buffer components. 100 reactions in a 20 μL total volume. Store at -5 to -30°C. Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
|
Tube | |
qPCR | |
Fast | |
SYBR Green | |
SYBR™ | |
UP (Ultra Pure) | |
Dry Ice | |
Gene Expression | |
100 Reactions | |
1 mL |
SYBR | |
High | |
AmpliTaq Fast DNA Polymerase, AmpliTaq Fast UP | |
7500 Fast System, 7900HT Fast System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode | |
ROX (Pre-mixed) | |
Real Time PCR SYBR Master Mix | |
1 x 1 mL | |
100 Reactions of 20 μL | |
2X | |
DNA (Genomic), cDNA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction