missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ SYBR™ Select Master Mix
Cost-effective solution for real-time PCR applications.
107.00 € - 3460.00 €
Specifications
Content And Storage | The 2x mix contains SYBR™ GreenER™ dye, AmpliTaq™ DNA Polymerase UP, dNTPs with dUTP/dTTP blend, heat-labile UDG, ROX passive reference dye and optimized buffer components. Contains 1 X 1 mL tube, sufficient for 100, 20 μL reactions (10 μL Master Mix per reaction). Store at 2–8°C. Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). See user guide for primer concentration recommendations for the StepOne and StepOnePlus systems. |
---|---|
Detection Method | SYBR |
Form | Liquid |
Format | Vial |
GC-Rich PCR Performance | High |
Includes: 2X mix contains SYBR GreenER dye, AmpliTaq DNA Polymerase UP, dNTPs with dUTP/dTTP blend, heat-labile UDG, ROX passive reference dye and optimized buffer components.
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13246519
|
Applied Biosystems™
4472903 |
1 x 1 mL |
107.00 €
100 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13256519
|
Applied Biosystems™
4472908 |
1 x 5 mL |
426.00 €
500 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13276519
|
Applied Biosystems™
4472918 |
2 x 5 mL |
786.00 €
1000 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13266519
|
Applied Biosystems™
4472919 |
5 x 5 mL |
1814.00 €
Pack of 2500 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13236529
|
Applied Biosystems™
4472920 |
10 x 5 mL |
3460.00 €
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13286519
|
Applied Biosystems™
4472913 |
1 x 50 mL |
3420.00 €
5000 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Specific - minimize primer-dimer and nonspecific amplification
- Reproducible and sensitive - consistent amplification across a wide dynamic range
- Bright - contains SYBR™ GreenER™ dye for maximum brightness
- Carry-over contamination control - contains heat-labile UDG
- Can be used in either standard or fast cycling mode
- SYBR™ Select Master Mix contains all the components needed for your real-time PCR reaction, except the template and primers, in a convenient 2X concentration premix
- The master mix includes AmpliTaq™ DNA Polymerase, UP, a highly purified DNA polymerase with a proprietary hot start mechanism that provides exceptional specificity
- SYBR™ Select Master Mix is specially formulated to provide robust results from 100ng to 0.1pg cDNA per reaction and can reliably detect a single copy of genomic DNA
- SYBR™ GreenER™ dye is less inhibitory to PCR than SYBR™ Green I dye resulting in brighter signals
- Heat-labile uracil-DNA glycosylase (UDG) is included for worry-free carryover contamination control
- SYBR™ Select Master Mix can be used in either standard or fast cycling mode and is compatible with all Applied Biosystems™ real-time PCR instruments except the 7900HT real-time PCR system
- It is also compatible with the Bio-Rad IQ™5, Roche LightCycler™ LC480, and Stratagene™ MX3005P™ systems
Order Info
Shipping Conditions: Wet Ice
Specifications
The 2x mix contains SYBR™ GreenER™ dye, AmpliTaq™ DNA Polymerase UP, dNTPs with dUTP/dTTP blend, heat-labile UDG, ROX passive reference dye and optimized buffer components. Contains 1 X 1 mL tube, sufficient for 100, 20 μL reactions (10 μL Master Mix per reaction). Store at 2–8°C. Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). See user guide for primer concentration recommendations for the StepOne and StepOnePlus systems. |
|
Liquid | |
High | |
AmpliTaq DNA Polymerase | |
SYBR GreenER | |
SYBR™ | |
UP (Ultra Pure) | |
Gene Expression | |
Built-In Hot Start |
SYBR | |
Vial | |
qPCR | |
Fast or Standard | |
ROX (Pre-mixed) | |
Real Time PCR SYBR Master Mix | |
100 Reactions of 20 μL | |
2X | |
DNA (Genomic), cDNA |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Applied Biosystems™ SYBR™ Select Master Mix