missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Universal Master Mix II, no UNG

TaqMan Universal Master Mix II is a convenient mix of components (except primers, probes, template, and water) necessary to perform a real-time polymerase chain reaction (PCR).

Brand:  Applied Biosystems™ 4440040

 View more versions of this product

Product Code. 10380155

  • 564.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Requires : Primers, probes, template, water
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Store at 2°C to 8°C in the dark.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).
High
AmpliTaq Gold DNA Polymerase
7300 System, 7500 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, ViiA™ 7 System
TaqMan™
UP (Ultra Pure)
Wet Ice
2X
DNA (Genomic), cDNA
Primer-probe
qPCR
Standard
ROX (Pre-mixed)
Universal Master Mix II
1 x 5 mL
Gene Expression, Genotyping, miRNA analysis
2 X
5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2791674 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number4440040,4440040_LITL18 PROMO
Lot Number2748832 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4440040,4440040_LITL18 PROMO
Lot Number2805007 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4440040,4440040_LITL18 PROMO
Lot Number2822266 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number4440040,4440040_LITL18 PROMO
Lot Number2907899 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number4440040,4440040_LITL18 PROMO
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Applied Biosystems™ TaqMan™ Universal Master Mix II, no UNG > 1 x 5 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.