missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Universal PCR Master Mix, no AmpErase™ UNG
TaqMan™ Universal PCR Master Mix, No AmpErase™ UNG is the ideal reagent solution when you need a Master Mix for multiple 5' nuclease DNA applications.
572.00 € - 5720.00 €
Specifications
Detection Method | Primer-probe |
---|---|
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | AmpliTaq Gold DNA Polymerase |
Reaction Speed | Standard |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10167154
|
Applied Biosystems™
4324020 |
10 x 5 mL, 2,000 reactions |
5720.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10157154
|
Applied Biosystems™
4324018 |
1 x 5 mL, 200 reactions |
572.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10462655
|
Applied Biosystems™
4326614 |
1 x 50 mL 2,000 reactions |
5560.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10437494
|
Applied Biosystems™
4364341 |
2 x 5 mL, 400 reactions |
1226.00 €
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10689835
|
Applied Biosystems™
4364343 |
5 x 5 mL, 1,000 reactions |
2940.00 €
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Contains AmpliTaq Gold™ DNA polymerase to provide a better yield and a more robust 5' nuclease assay than AmpliTaq™ DNA polymerase
- Includes a passive internal reference based on proprietary ROX™ dye, for superb precision on Applied Biosystems™ real-time PCR instruments
- Buffer enhancements guarantee performance and reliability in all applications, even with G/C-rich sequences
- Contains dUTP in the dNTP mix to protect against carry-over contamination, when used with AmpErase™ UNG
- Rapid assay development guidelines are provided to minimize optimization time
Order Info
Guaranteed minimum shelf life is 60 days. Shipping Conditions: Wet Ice.
Specifications
Primer-probe | |
qPCR | |
Standard | |
ROX (Pre-mixed) | |
Universal PCR Master Mix | |
Gene Expression, Genotyping, miRNA analysis |
High | |
AmpliTaq Gold DNA Polymerase | |
7000 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, GeneAmp 9600, StepOne™, Standard Mode, StepOnePlus™, Standard Mode | |
TaqMan™ | |
Wet Ice | |
DNA (Genomic), cDNA |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title