Learn More
Stuart™ Aquatron™ deionizer Housing and Accessories
Stuart™ Aquatron™ deionizer Housing and Accessories provide good, quality water at an affordable price
Brand: Stuart™ 90269-78
This item is not returnable.
View return policy
Description
- Output up to 60 litres/hour
- Low cost disposable cartridges
- Colour change indicates resin condition at a glance
- Free standing or wall mountable
- Ideal as pre-treatment for Aquatron™ water stills
- A simple, portable deionizer giving good quality water at an affordable price
- The disposable ion exchange cartridges slowly change colour from green to blue as they are exhausted giving at a glance indication of the resin condition
- No need for conductivity meters, batteries or mains electrical power
- Supplied complete with flexible hose for connection to a tap and a stopcock to control the flow rate
- Can be linked in series with the Aquatron™ filter either by a flexible hose or the rigid coupling (ALC) available as an accessory
- Ideal as a pre-treatment for Aquatron™ water stills to prevent scale build up and improve output quality
- Housing and ion exchange cartridges must be ordered separately
Specifications
Aquatron Filter | |
Wall Bracket | |
Single |