missing translation for 'onlineSavingsMsg'
Learn More

Stuart™ Aquatron™ deionizer Housing and Accessories

Stuart™ Aquatron™ deionizer Housing and Accessories provide good, quality water at an affordable price

23.70 € - 620.00 €

Specifications

For Use With (Equipment) Aquatron Filter
View More Specs

Products 4
Product Code Brand Type Features Price Quantity & Availability  
Product Code Brand Type Features Price Quantity & Availability  
16150921
View Documents
Stuart™
80244-94
Deionizer Housing Double Capacity
620.00 €
Each
Estimated Shipment: 20-01-2025
to see stock.
Add to basket
 
16170921
View Documents
Stuart™
80244-96
Deionizer Housing Single Capacity
321.00 €
Each
In Stock
to see stock.
Add to basket
 
16160921
View Documents
Stuart™
90269-78
Wall Bracket Single
23.70 €
Each
In Stock
to see stock.
Add to basket
 
16140921
View Documents
Stuart™
80244-95
Wall Bracket Double
33.15 €
Each
In Stock
to see stock.
Add to basket
 
Description

Description

  • Output up to 60 litres/hour
  • Low cost disposable cartridges
  • Colour change indicates resin condition at a glance
  • Free standing or wall mountable
  • Ideal as pre-treatment for Aquatron™ water stills
  • A simple, portable deionizer giving good quality water at an affordable price
  • The disposable ion exchange cartridges slowly change colour from green to blue as they are exhausted giving at a glance indication of the resin condition
  • No need for conductivity meters, batteries or mains electrical power
  • Supplied complete with flexible hose for connection to a tap and a stopcock to control the flow rate
  • Can be linked in series with the Aquatron™ filter either by a flexible hose or the rigid coupling (ALC) available as an accessory
  • Ideal as a pre-treatment for Aquatron™ water stills to prevent scale build up and improve output quality
  • Housing and ion exchange cartridges must be ordered separately
Specifications

Specifications

Aquatron Filter
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.