missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ AR-GLAS™ Stirring Rods
Constructed of AR-GLAS™. Tubes are fused closely at both ends.
34.05 € - 80.40 €
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10283821
|
BRAND™
135810 |
200 mm |
34.05 €
Pack of 50 |
In Stock
Log in to see stock. |
|||||
10597641
|
BRAND™
135820 |
250 mm |
71.00 €
Pack of 50 |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
10014321
|
BRAND™
135825 |
300 mm |
80.40 €
Pack of 50 |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
Description
- Constructed of AR-GLAS™
- Tubes are fused closely at both ends.
- Supplied in packs of 50
Specifications
Glass | |
50/Pk. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title