Learn More
Thermo Scientific™ Arctic Express™ Dual Storage Systems
Store and ship samples with Arctic Express dual storage systems, which feature hard-shell shipping containers.
Brand:  Thermo Scientific™ CK50922
 View more versions of this product
Product Code. 10699463
  • 3316.50 € / Each
Estimated Shipment: 25-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Protect nonhazardous samples during shipment and storage using Thermo Scientific™ Arctic Express™ Storage Systems. Featuring excellent flexibility and safety features, these systems can be used as dry shippers or laboratory cryo vessels.
Features:
  • When filled with liquid nitrogen and placed in the laboratory, unit can be used as a “can and cane” storage system indefinitely with static holding times of up to 125 days.
  • Aluminum vessels come with lockable lid and hold six numbered, color-coded, stainless-steel cans (included), which accommodate canes that hold ampules.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Manual Fill LN2 Dewar
1
21 shipper/50 lab
3.8 in.
9.7 cm
28 L
Arctic Express Dual 28
18.2 in.
46.2 cm
630 x 2mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products