Learn More
Thermo Scientific™ Arctic Express™ Dual Storage Systems
Store and ship samples with Arctic Express dual storage systems, which feature hard-shell shipping containers.
2401.95 € - 3316.50 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10338253
|
Thermo Scientific™
CK50920 |
10 L |
2401.95 €
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
|||||
10699463
|
Thermo Scientific™
CK50922 |
28 L |
3316.50 €
Each |
Estimated Shipment: 14-03-2025
Log in to see stock. |
|||||
Description
Protect nonhazardous samples during shipment and storage using Thermo Scientific™ Arctic Express™ Storage Systems. Featuring excellent flexibility and safety features, these systems can be used as dry shippers or laboratory cryo vessels.
Features:- When filled with liquid nitrogen and placed in the laboratory, unit can be used as a “can and cane” storage system indefinitely with static holding times of up to 125 days.
- Aluminum vessels come with lockable lid and hold six numbered, color-coded, stainless-steel cans (included), which accommodate canes that hold ampules.
- Warranty varies by country, please contact us for details
Specifications
Manual Fill LN2 Dewar | |
1 |