Learn More
Sartorius™ arium™ bagtanks
For secure storage and protection of pure water from secondary contamination.
Brand:  Sartorius™ H2O-AOV-100
2140.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 14851372
  • 2195.00 € / Each
Explore more special offers
Description
Description
  • Integrated venting filter with check valve securely protects against CO2 contamination
  • Optional rollers provide flexibility
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 L
Pure water is securely protected from secondary contamination
Stainless Steel, Plastic
Sterile
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Sartorius™ arium™ bagtanks >