Learn More
Sartorius Arium™ Smart Station UP
GREENER_CHOICE
Arium™ Smart Station flexible remote dispenser for ultrapure water
Brand:  Sartorius H2O-ARST-UP-B
 View more versions of this product
Product Code. 17274233
  • 2350.00 € / 1 unit
Explore more special offers
Description
Description
  • Height adjustable display/dispense station for dispensing Type 1 ultrapure water
  • Compact design for easy and space-saving integration into lab environment
  • Intuitive touch-activated color display with direct access to important functions
  • Flexible and stepless height adjustment to fill different sized containers and beakers
  • Ergonomic design can be optimized for right or left hand operation
  • Accurate volume dispense for fast and precise buffer and sample prep
  • Adjustable, splash-free flow rate from drop-by-drop to 2 L/min
Equipment supplied:
  • Display/dispense unit with stand
  • Power supply including:
    • IEC connector
    • Connection set
For intended use with:
  • Arium™ Bagtank
  • Arium™ Pro
  • Arium™ Comfort
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 mL to 50 L
Ultrapure (Type 1)
Touch display
Ultrapure
Smart Station Wall-Mounted