missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Arium™ Smart Station UP
Arium™ Smart Station flexible remote dispenser for ultrapure water
2350.00 €
Specifications
Capacity (Metric) | 50 mL to 50 L |
---|---|
Feedwater Source | Ultrapure |
Product Water Type | Ultrapure (Type 1) |
Display | Touch display |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
17284233
|
Sartorius
H2O-ARST-UP-T |
Smart Station Benchtop |
2350.00 €
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17274233
|
Sartorius
H2O-ARST-UP-B |
Smart Station Wall-Mounted |
2350.00 €
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Height adjustable display/dispense station for dispensing Type 1 ultrapure water
- Compact design for easy and space-saving integration into lab environment
- Intuitive touch-activated color display with direct access to important functions
- Flexible and stepless height adjustment to fill different sized containers and beakers
- Ergonomic design can be optimized for right or left hand operation
- Accurate volume dispense for fast and precise buffer and sample prep
- Adjustable, splash-free flow rate from drop-by-drop to 2 L/min
Equipment supplied:
- Display/dispense unit with stand
- Power supply including:
- IEC connector
- Connection set
For intended use with:
- Arium™ Bagtank
- Arium™ Pro
- Arium™ Comfort
Specifications
50 mL to 50 L | |
Ultrapure | |
Ultrapure (Type 1) | |
Touch display |