Learn More
Abeba™ Arrow 35600 Shoes
Antistatic with heel patch microfiber. Sanitized™ treated.
Brand:  Abeba™ 35600/46
 View more versions of this product
Product Code. 15680188
  • 86.15 € / 1 pair
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, FO SRA
  • Smooth leather, perforated
  • White
  • Slip resistance PU outsole sole
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • Microfiber heel patch
  • Leather lining
Specifications
Specifications
CE EN ISO 20347:2012 OB A, FO SRA
Leather
Slip-resistant
35600
White
46
Polyurethane