Learn More
Thermo Scientific™ ART™ Barrier Pipette Tips in Hinged Racks
Perform pipetting accurately and consistently with these barrier pipette tips in hinged racks, which seal when exposed to potential contaminants. ART hinged racks are available in low retention, micropoint, extended length and other specialty styles.
113.00 € - 177.00 €
Specifications
Filtered | Filtered |
---|---|
Sterility | Sterile |
Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
For Use With (Equipment) | Universal Fit |
Product Line | ART |
Product Code | Brand | Volume (Metric) | Model | Format | Tip Style | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Model | Format | Tip Style | Price | Quantity & Availability | |||
10217184
![]() |
Thermo Scientific™
2139-HR |
10 μL | ART 10 | Hinged Rack | Universal Fit |
147.00 €
Pack of 960 |
In Stock
Log in to see stock. ![]() |
|||
11942689
![]() |
Thermo Scientific™
2139-05-HR |
10 μL | ART 10 | Hinged Rack | Low Retention |
123.02 €
Pack of 960 |
In Stock
Log in to see stock. ![]() |
|||
10706815
![]() |
Thermo Scientific™
2149E-HR |
10 μL | ART 10 Ultra-micro | Hinged Rack | Ultra-micro |
115.44 €
Pack of 960 |
In Stock
Log in to see stock. |
|||
10032874
![]() |
Thermo Scientific™
2149E-05-HR |
10 μL | ART 20E Ultra-micro | Hinged Rack | Low Retention |
124.00 €
Pack of 960 |
In Stock
Log in to see stock. ![]() |
|||
10401755
![]() |
Thermo Scientific™
2149P-HR |
20 μL | ART 20P | Hinged Rack | Universal Fit |
160.00 €
Pack of 960 |
In Stock
Log in to see stock. ![]() |
|||
10276944
![]() |
Thermo Scientific™
2149P-05-HR |
20 μL | ART 20P | Hinged Rack | Low Retention |
177.00 €
Pack of 960 |
In Stock
Log in to see stock. ![]() |
|||
10777385
![]() |
Thermo Scientific™
2065-HR |
100 μL | ART 100 | Hinged Rack | Universal Fit |
113.00 €
Pack of 960 |
In Stock
Log in to see stock. |
|||
10746625
![]() |
Thermo Scientific™
2065E-HR |
100 μL | ART 100E | Hinged Rack | Universal Fit |
133.00 €
Pack of 960 |
In Stock
Log in to see stock. ![]() |
|||
12661480
![]() |
Thermo Scientific™
2065E-05-HR |
100 μL | ART 100E | Hinged Rack | Low Retention |
155.00 €
Pack of 960 |
In Stock
Log in to see stock. ![]() |
|||
10431375
![]() |
Thermo Scientific™
2069-HR |
200 μL | ART 200 | Hinged Rack | Universal Fit |
159.00 €
Pack of 960 |
In Stock
Log in to see stock. ![]() |
|||
10768735
![]() |
Thermo Scientific™
2069-05-HR |
200 μL | ART 200 | Hinged Rack | Low Retention |
124.84 €
Pack of 960 |
In Stock
Log in to see stock. ![]() |
|||
15173606
![]() |
Thermo Scientific™
2070-HR |
300 μL | ART Barrier | Rack | Universal Fit |
144.00 €
Pack of 960 |
Estimated Shipment: 24-09-2025
Log in to see stock. |
|||
15193606
![]() |
Thermo Scientific™
2070-05-HR |
300 μL | ART Barrier | Rack | Low Retention |
155.00 €
Pack of 960 |
Estimated Shipment: 30-10-2025
Log in to see stock. |
|||
10564914
![]() |
Thermo Scientific™
2179-HR |
1000 μL | ART Barrier | Hinged Rack | Universal Fit |
164.00 €
Pack of 768 |
In Stock
Log in to see stock. |
|||
Description
Unlike standard filter tips, Thermo Scientific™ ART™ Barrier Hinged Rack Pipette Tips seal when exposed to potential contaminants. The pipette tips from the reload inserts fit securely into the environmentally friendly, reusable hinged racks and the covers may be used as hinged or lift-off style. The locking clasp prevents contamination by keeping remaining tips sterile and offers an easy, one-handed operation. ART hinged racks are available in low retention, micropoint, extended length, and other specialty styles.
Features:- Stackable hinged racks save space in your lab
- Sterile tips are DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
Recommended for:
- Liquid Handling
- Molecular Biology
- PCR
- Biotechnology
Specifications
Filtered | |
DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free | |
ART |
Sterile | |
Universal Fit | |
Sterile and PCR Ready |