missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ART™ Barrier Specialty Pipette tips
GREENER_CHOICE

Perform pipetting accurately and consistently with these barrier extended length pipette tips, which seal when exposed to potential contaminants.

Brand:  Thermo Scientific™ 2279

98.40 EUR valid until 2025-05-31
Use promo code "25076" to get your promotional price.


 View more versions of this product

Product Code. 10045460

  • 176.00 € / Pack of 800
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Perform applications consistently and accurately by using Thermo Scientific™ ART™ Barrier Specialty Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier specialty tips are designed to aid in pipetting for a variety of applications. Extended length, Thermo Scientific™ REACH™ tips provide an extra level of contamination control.

Features:

  • Sterile tips are certified free from RNase, DNase and pyrogen
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard

Designed to Aid in Ppipetteting for a Variety of Applications:

Extended length: to prevent contamination by keeping the pipettete shaft from entering the sample vessel

Gel loading: for loading acrylamide or agarose gels

Wide orifice genomic: for genomic DNA, fragile cell lines and other viscous materials

Solvent safe: for acids, bases and aggressive organic solvents

Compatible with:

SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettes

Ergonomic pipette tips are designed for Gilson Pipetman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes

TRUSTED_SUSTAINABILITY
Specifications

Specifications

ART 1000, Filtered, Sterile, Lift-off Lid Rack
Filtered
ART 1000
7.9cm
Lift-off Lid Rack
Gilson™ Pipetman™, Thermo Scientific™ Finnpipette™ Focus
Sterile and PCR Ready
1000 μL
Sterile
3.13 in.
DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Ergonomic
ART
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1357655 Certificate TypeCertificate of Compliance Date13/05/2022 Catalog Number2279
Lot Number1345968 Certificate TypeCertificate of Compliance Date29/01/2022 Catalog Number2279
Lot Number1337926 Certificate TypeCertificate of Compliance Date18/01/2022 Catalog Number2279
Lot Number1337925 Certificate TypeCertificate of Compliance Date04/12/2021 Catalog Number2279
Lot Number1337924 Certificate TypeCertificate of Compliance Date30/11/2021 Catalog Number2279
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ ART™ Barrier Specialty Pipette tips > ART™ Barrier Extended Length Pipette Tips; Volume: 1000μL; Tip Model: ART 1000 soft fit; Tip style: MicroPoint; Sterility: Sterile; Unit Size: Case of 4 x Packs of 8 x Racks of 100 tips (3200 tips in total)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.