Learn More
Thermo Scientific™ ART™ Barrier Specialty Pipette tips

Perform pipetting accurately and consistently with these barrier extended length pipette tips, which seal when exposed to potential contaminants.
121.00 € - 176.00 €
Specifications
Filtered | Filtered |
---|---|
Sterility | Sterile |
Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
Format | Lift-off Lid Rack |
Product Line | ART |
Product Code | Brand | Volume (Metric) | Model | Tip Style | Fit for Purpose | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Model | Tip Style | Fit for Purpose | Price | Quantity & Availability | |||
10399360
![]() |
Thermo Scientific™
2770 |
200 μL | ART 200U | Ergonomic | Sterile and PCR Ready |
121.00 €
Pack of 960 |
In Stock
Log in to see stock. |
|||
10142721
![]() |
Thermo Scientific™
2780 |
50 μL | ART 50U | Ergonomic | Sterile and PCR Ready |
121.00 €
Pack of 960 |
In Stock
Log in to see stock. |
|||
10045460
![]() |
Thermo Scientific™
2279 |
1000 μL | ART 1000 | Ergonomic | Sterile and PCR Ready |
176.00 €
Pack of 800 |
In Stock
Log in to see stock. ![]() |
|||
10078950
![]() |
Thermo Scientific™
2140 |
10 μL | ART 10 REACH | Extended Length | Sterile and PCR Ready |
128.88 €
Pack of 960 |
In Stock
Log in to see stock. ![]() |
|||
10008970
![]() |
Thermo Scientific™
2149 |
0.5 to 20 μL | ART 20E | Micro Point | Sterile and PCR Ready |
122.00 €
Pack of 960 |
In Stock
Log in to see stock. |
|||
Description
Perform applications consistently and accurately by using Thermo Scientific™ ART™ Barrier Specialty Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier specialty tips are designed to aid in pipetting for a variety of applications. Extended length, Thermo Scientific™ REACH™ tips provide an extra level of contamination control.
Features:
- Sterile tips are DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
- Available in sterile, nonsterile and in various packaging options
- Manufactured according to the industry's highest quality standard
Designed to Aid in Pipetting for a Variety of Applications:
Extended length: to prevent contamination by keeping the pipette shaft from entering the sample vessel
Gel loading: for loading acrylamide or agarose gels
Wide orifice genomic: for genomic DNA, fragile cell lines and other viscous materials
Solvent safe: for acids, bases and aggressive organic solvents
Compatible with:
SoftFit™ pipette tips provide precise and consistent pipetting with multichannel pipettes
Ergonomic pipette tips are designed for Gilson Pipetman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes
Specifications
Filtered | |
DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free | |
ART |
Sterile | |
Lift-off Lid Rack | |
Sterile and PCR Ready |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.